Learn More
Invitrogen™ Cofilin Recombinant Rabbit Monoclonal Antibody (24GB4775)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557363
 View more versions of this product
Product Code. 30275590
  • 564.00 € / 100µL
Description
Cofilin Recombinant Monoclonal Antibody for Western Blot, Flow
Cofilin binds to F-actin and exhibits pH-sensitive F-actin depolymerizing activity. Regulates actin cytoskeleton dynamics. Important for normal progress through mitosis and normal cytokinesis. Plays a role in the regulation of cell morphology and cytoskeletal organization. Required for the up-regulation of atypical chemokine receptor ACKR2 from endosomal compartment to cell membrane, increasing its efficiency in chemokine uptake and degradation. Required for neural tube morphogenesis and neural crest cell migration.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cofilin
Recombinant Monoclonal
Unconjugated
CFL1
18 kDa phosphoprotein; AA959946; CFL; Cfl1; CFL-1; Cof; COF1; cofilin; cofilin 1; cofilin 1 (non-muscle); cofilin 1, non-muscle; cofilin, non-muscle isoform; Cofilin1; Cofilin-1; cofilin-1-like protein; epididymis secretory protein Li 15; HEL-S-15; n-cofilin; p18
Rabbit
Affinity chromatography
RUO
1072, 12631, 29271
-20°C
Liquid
Flow Cytometry, Western Blot
24GB4775
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P18760, P23528, P45592
CFL1
A synthesized peptide derived from human Cofilin (aa 116-129).
100 μL
Primary
Human, Mouse, Rat