missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COL12A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104546
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Collagen XII alpha 1.
This gene encodes the alpha chain of type XII collagen, a member of the FACIT (fibril-associated collagens with interrupted triple helices) collagen family. Type XII collagen is a homotrimer found in association with type I collagen, an association that is thought to modify the interactions between collagen I fibrils and the surrounding matrix. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq, Jul 2008].
Specifications
| COL12A1 | |
| Polyclonal | |
| Unconjugated | |
| COL12A1 | |
| BA209D8.1; BTHLM2; COL12A1; COL12A1L; collagen alpha-1(XII) chain; collagen type XII alpha 1 chain; collagen type XII proteoglycan; collagen, type XII, alpha 1; DJ234P15.1; UCMD2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1303 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q99715 | |
| COL12A1 | |
| A synthesized peptide derived from human COL12A1(Accession Q99715), corresponding to amino acid residues Y1069-V1119. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction