missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Procollagen I N-Peptide Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA535379
This item is not returnable.
View return policy
Descripción
This antibody is predicted to react with most mammals, birds, amphibians and fish based on 100% sequence homology. This antibody contains enough material to conduct 10 mini-Western blots.
Procollagen type I N-terminal propeptide (P1NP) is a specific marker of bone formation, reflecting the synthesis of type I collagen, which constitutes the major part of the bone matrix. P1NP is derived during the conversion of procollagen to collagen, produced by osteoblasts during bone formation. Its primary clinical application lies in the diagnosis and monitoring of metabolic bone diseases, particularly osteoporosis. In osteoporotic patients, P1NP levels can predict bone mineral density (BMD) responses to treatments like teriparatide and romosozumab. Low serum levels of P1NP are indicative of a high risk of BMD loss, providing a valuable tool for premenopausal women with systemic lupus erythematosus (SLE) and other conditions affecting bone health.
Especificaciones
| Procollagen I N-Peptide | |
| Polyclonal | |
| Unconjugated | |
| COL1A1 | |
| P1NP; Procollagen I N | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1277, 12842, 29393 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.15 mg/mL | |
| PBS with no preservative; pH 7.5 | |
| P02452, P02454, P11087 | |
| COL1A1 | |
| Synthetic peptide corresponding to amino acid residues specific to human collagen 1, alpha 1 propeptide conjugated to KLH. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido