Learn More
Invitrogen™ COL1A1 Recombinant Rabbit Monoclonal Antibody (K01_2U53)
Rabbit Recombinant Monoclonal Antibody
287.00 € - 498.00 €
Specifications
| Antigen | COL1A1 |
|---|---|
| Clone | K01_2U53 |
| Concentration | 0.60 mg/mL |
| Content And Storage | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
| Applications | Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30828583
|
Invitrogen™
MA564286 |
100 μL |
498.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30826646
|
Invitrogen™
MA565745 |
50 μL |
287.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: KAHDGGRYYR ADDANVVRDR DLEVDTTLKS LSQQIENIRS PEGSRKNPAR TCRDLKMCHS DWKSGEYWID PNQGCNLDAI KVFCNMETGE TCVYPTQPSV AQKNWYISKN PKDKRHVWFG ESMTDGFQFE YGGQGSDPAD VAIQLTFLRL MSTEASQNIT YHCKNSVAYM DQQTGNLKKA LLLQGSNEIE IRAEGNSRFT YSVTVDGCTS HTGAWGKTVI EYKTTKTSRL PIIDVAPLDV GAPDQE Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%, Pig - 98%, Cynomolgus monkey - 100%.
The protein encoded by this gene is the pro-alpha1 chain of type I collagen whose triple helix comprises two alpha1 chains and one alpha2 chain. Type I is a fibril-forming collagen found in most connective tissues and is abundant in bone, cornea, dermis and tendon. Mutations in this gene are associated with osteogenesis imperfecta types I-IV, Ehlers-Danlos syndrome type VIIA, Ehlers-Danlos syndrome Classical type, Caffey Disease and idiopathic osteoporosis. Reciprocal translocations between chromosomes 17 and 22, where this gene and the gene for platelet-derived growth factor beta are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans, resulting from unregulated expression of the growth factor. Two transcripts, resulting from the use of alternate polyadenylation signals, have been identified for this gene.Specifications
| COL1A1 | |
| 0.60 mg/mL | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4 | |
| P02452 | |
| 1277 | |
| Recombinant protein of human Collagen I (aa 1208-1453). | |
| Antibody |
| K01_2U53 | |
| Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| COL1A1 | |
| alpha-1 type 1 collagen; alpha-1 type I collagen; alpha1(I) procollagen; COL1A; Col1a1; Col1a-1; Cola1; Cola-1; COLIA1; Collagen 1; collagen alpha 1 chain type I; collagen alpha-1(I) chain; collagen alpha-1(I) chain preproprotein; Collagen I; collagen of skin, tendon and bone, alpha-1 chain; collagen type I alpha 1; collagen, type 1, alpha 1; collagen, type I, alpha 1; Collagen1A1; EDSC; Mov13; Mov-13; OI1; OI2; OI3; OI4; pro-alpha-1 collagen type 1; procollagen type I, alpha 1; procollagen, type 1, alpha 1; procollagen, type I, alpha 1; Type I Collagen; type I proalpha 1; type I procollagen alpha 1 chain; type I-alpha 1 collagen | |
| COL1A1 | |
| Primary | |
| Protein A/G |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.