missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COLEC12 Monoclonal Antibody (388305)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561197
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene encodes a member of the C-lectin family, proteins that possess collagen-like sequences and carbohydrate recognition domains. This protein is a scavenger receptor, a cell surface glycoprotein that can bind to carbohydrate antigens on microorganisms facilitating their recognition and removal. In addition, these receptors can recognize oxidized phospholipids so they may also participate in removing oxidatively damaged or apoptotic cells.
Specifications
| COLEC12 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q5KU26 | |
| Colec12 | |
| Mouse myeloma cell line NS0-derived recombinant human CL-P1/COLEC12 isoform 1, Leu57-Leu742. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 388305 | |
| Unconjugated | |
| Colec12 | |
| CLP1; CL-P1; COLEC12; Collectin placenta protein 1; collectin subfamily member 12; collectin sub-family member 12; collectin-12; hCL-P1; NSR2; Nurse cell scavenger receptor 2; Scara4; Scavenger receptor class A member 4; scavenger receptor class A, member 4; scavenger receptor with C-type lectin; Srcl | |
| Rat | |
| Protein A/G | |
| RUO | |
| 81035 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction