missing translation for 'onlineSavingsMsg'
Learn More

Colistin sulfate, Thermo Scientific™

Colistin Sulfate salt (2:5), CAS # 1264-72-8, (synonym: Polymyxin E Complex), is a complex mixture of antibiotics, with Colistin A and B being the primary components. It has anti-bacterial activity against Gram-positive and Gram-negative bacteria and may also be used as a neuromuscular blocker.

Brand:  Thermo Scientific Alfa Aesar J60915.03


Quantity 1g

Product Code. 15463599

  • ONLINE EXCLUSIVE PRICE
    181.00 €
    177.00 € / 1g
    Save 4.00 €
    2% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General description
• Colistin Sulfate salt is a stable, cationic, water-soluble, white powder composed of over 30 components
• Originally isolated from B. polymyxa, it inhibits the growth of the Gram-negative (e.g. E. coli, P. aeruginosa, P. fluorescens, and S. enterica), and Gram-positive bacteria (e.g. L. lactis, P. polymyxa, P. acidilactici, and S. aureus) by permeabilizing the bacterial cell membrane
Applications
• Colistin Sulfate has potent anti-endotoxin activity rendered through its ability to bind lippopolysaccharides that make up bacterial endotoxins
• This compound is often used in the management of multi-drug resistant Gram-negative infections
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1264-72-8
1g
UN2811
Polymyxin E
Colistin sulfate
White
MFCD00146495
14,2479
Soluble in water
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Colistin sulphate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3

Hazard Statement
  • H301-Toxic if swallowed.

Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

Supplemental information
  • MIXTURE LIST-Contains : Colistin, sulphate

SDS Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.