CAS RN 1264-72-8
CAS RN 1264-72-8
IUPAC Name:
N-[(1S)-3-amino-1-{[(1S,2R)-1-{[(1S)-3-amino-1-{[(3S,6S,9S,12S,15R,18S,21S)-6,9,18-tris(2-aminoethyl)-3-[(1R)-1-hydroxyethyl]-12,15-bis(2-methylpropyl)-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptaazacyclotricosan-21-yl]carbamoyl}propyl]carbamoyl}-2-hydroxypropyl]carbamoyl}propyl]-6-methyloctanamide; sulfuric acid
Synonyms:
colistin
Molecular Weight (g/mol):
1267.55
Molecular Formula:
C53H102N16O17S
InChi Key:
ZJIWRHLZXQPFAD-LRYSGCCDSA-N
SMILES:
OS(O)(=O)=O.CCC(C)CCCCC(=O)N[C@@H](CCN)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](CCN)C(=O)N[C@H]1CCNC(=O)[C@@H](NC(=O)[C@H](CCN)NC(=O)[C@H](CCN)NC(=O)[C@H](CC(C)C)NC(=O)[C@@H](CC(C)C)NC(=O)[C@H](CCN)NC1=O)[C@@H](C)O
1
–
6
of
6
results
1
Thermo Scientific Chemicals Colistin sulfate salt
CAS: 1264-72-8
3
Colistin, sulfate salt, MP Biomedicals™
CAS: 1264-72-8
5
Colistin sulfate 100 μg/mL in Water, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
6
Colistin sulfate, MedChemExpress
MedChemExpress Colistin sulfate is a polypeptide antibiotic which inhibits gram-negative bacteria by binding to lipopolysaccharides and phospholipids in the outer cell membrane of gram-negative bacteria.