missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen I alpha 1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77458SS
This item is not returnable.
View return policy
Description
Collagen I alpha 1 Polyclonal specifically detects Collagen I alpha 1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Western Blot (Negative).
Specifications
| Collagen I alpha 1 | |
| Polyclonal | |
| Unconjugated | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| Alpha-1 type I collagen, COL-IA1, Collagen 1, Collagen 1 alpha 1, collagen alpha 1 chain type I, collagen alpha-1(I) chain, collagen of skin, tendon and bone, alpha-1 chain, Collagen Type I Alpha 1 Chain, collagen, type I, alpha 1, Collagen1, Collagen-1, EDSARTH1, OI4, pro-alpha-1 collagen type 1, Type I Procollagen Alpha 1 Chain | |
| Rabbit | |
| 139 kDa | |
| 0.025 mL | |
| Cell Biology, Cellular Markers, Extracellular Matrix, Signal Transduction, Stem Cells | |
| 1277 | |
| Human, Mouse | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1.12 mg/mL | |
| Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 10-20 ug/ml, Immunohistochemistry-Paraffin 1:200, Western Blot (Negative) | |
| P02452 | |
| COL1A1 | |
| This Collagen I alpha 1 antibody was raised against a synthetic peptide made to the C-terminal portion of the human Collagen I protein (between residues 1300-1350) [UniProt P02452] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Mouse. Predicted reactivity in Human. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction