Learn More
Collagen I Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 195 publications
Brand:  Novus Biologicals NB600-408-0.1mg
 View more versions of this product
Product Code. 18729023
  • 522.00 € / 0.10mg
Description
Collagen I Polyclonal specifically detects Collagen I in Human, Mouse, Rat, Porcine, Bacteria, Bovine, Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, SDS-Page, Fluorophore-linked immunosorbent assay, Microarray.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Collagen I alpha 1
Polyclonal
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Alpha-1 type I collagen, Collagen 1, Collagen 1 alpha 1, collagen alpha 1 chain type I, collagen alpha-1(I) chain, collagen of skin, tendon and bone, alpha-1 chain, collagen, type I, alpha 1, Collagen1, Collagen-1, OI4, pro-alpha-1 collagen type 1
Rabbit
139 kDa
0.1 mg
Cancer, Cell Cycle and Replication, Cytoskeleton Markers, Extracellular Matrix
1277
Human
IgG
Western Blot
1 mg/mL
Western Blot 1:1000, ELISA 1:5000-1:50000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:10-1:500, Immunocytochemistry 1:1600
P02452, P02453
COL1A1
Collagen Type I from human and bovine placenta by limited pepsin digestion and selective salt precipitation.
Affinity Purified
RUO
Primary
Typically less than 1% cross-reactivity against other types of collagens was detected by ELISA against purified standards. Some class-specific anti-collagens may be specific for three-dimensional epitopes which may result in diminished reactivity with denatured collagen or formalin-fixed, paraffin embedded tissues. This antibody reacts with most mammalian Type I collagens and has negligible cross-reactivity with Type II, III, IV, V or VI collagens. Non-specific cross-reaction of anti-collagen antibodies with other human serum proteins or non-collagen extracellular matrix proteins is negligible.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.