missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Collagen I Native Protein

Product Code. 18729423 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18729423 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18729423 Supplier Novus Biologicals™ Supplier No. NBP197266
  • ONLINE EXCLUSIVE PRICE
    592.00 €
    559.65 € / 0.5mg
    Save 32.35 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

Highly purified. Generating reliable and reproducible results. Applications: Western Blot, ELISA, Immunohistochemistry

Native COL1A1. Source: Human Placenta (Accession #: P02452) The Collagen I alpha 1 Native Protein has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P02452, P02452, P02452, P02452
Concentration 1.0 mg/mL
For Use With (Application) ELISA, Immunohistochemistry, Western Blot
Formulation 0.5 M Acetic Acid
Gene ID (Entrez) 1277
Name Collagen I alpha 1 protein
Quantity 0.5 mg
Source Human serum
Storage Requirements Store at 4 C. Do not freeze.
Gene Symbol COL1A1
Cross Reactivity Human
Research Category Cell Biology, Cellular Markers, Extracellular Matrix, Signal Transduction, Stem Cells
Purity or Quality Grade Multi-step
Protein Collagen I alpha 1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.