missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen IV alpha5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92585-0.02ml
This item is not returnable.
View return policy
Description
Collagen IV alpha5 Polyclonal antibody specifically detects Collagen IV alpha5 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Collagen IV alpha5 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:500 - 1:1000 | |
| alpha-5 polypeptide, Alport syndrome, CA54, collagen of basement membrane, alpha-5 chain, collagen, type IV, alpha 5, dA149D17.3, dA24A23.1, MGC167109, MGC42377 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1462-1691 of human COL4A5 (NP_203699.1). SSVAHGFLITRHSQTTDAPQCPQGTLQVYEGFSLLYVQGNKRAHGQDLGTAGSCLRRFSTMPFMFCNINNVCNFASRNDYSYWLSTPEPMPMSMQPLKGQSIQPFISRCAVCEAPAVVIAVHSQTIQIPHCPQGWDSLWIGYSFMMHTSAGAEGSGQALASPGSCLEEFRSAPFIECHGRGTCNYYANSYSFWLATVDVSDMFSKPQSETLKAGDLRTRISRCQVCMKRT | |
| 0.02 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 1287 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction