missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen IV alpha5 Antibody (COL4A5/9396R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-23832-20ug
This item is not returnable.
View return policy
Description
Collagen IV alpha5 Monoclonal antibody specifically detects Collagen IV alpha5 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Collagen IV alpha5 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| alpha-5 polypeptide, Alport syndrome, CA54, collagen of basement membrane, alpha-5 chain, collagen, type IV, alpha 5, dA149D17.3, dA24A23.1, MGC167109, MGC42377 | |
| Recombinant fragment (around aa1400-1685) of human Collagen IV alpha5 protein (exact sequence is proprietary) | |
| 20 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| COL4A5/9396R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 1287 | |
| Store at 4°C. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction