missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen IV Antibody (COL4/9395R), DyLight 650, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-24322C
This item is not returnable.
View return policy
Description
Collagen IV Monoclonal antibody specifically detects Collagen IV in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Collagen IV | |
| Monoclonal | |
| DyLight 650 | |
| BSVD;BSVD1;COL4A1s;Collagen alpha-1(IV) chain;PADMAL;RATOR | |
| Recombinant fragment (around aa1400-1670) of human Collagen IV protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Extracellular Matrix, Stem Cell Markers | |
| 1282 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| COL4/9395R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction