missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen IV Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB120-6586
This item is not returnable.
View return policy
Description
Collagen IV Polyclonal specifically detects Collagen IV in Human, Mouse, Rat, Bovine, Feline samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating, Knockout Validated, Super-Resolution Microscopy.
Specifications
| Collagen IV alpha 1 | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| alpha-1 chain, arresten, COL4A1 NC1 domain, collagen alpha-1(IV) chain, collagen IV, alpha-1 polypeptide, collagen, type IV, alpha 1, Collagen-4, EC 3.4.23, EC 6.3.1.2 | |
| Rabbit | |
| 160.61 kDa | |
| 0.1 mg | |
| Cytoskeleton Markers, Extracellular Matrix, Stem Cell Markers | |
| 1282 | |
| Human, Mouse, Rat, Bovine, Feline, Mammalia | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry (Free Floating), Immunoassay, CyTOF | |
| 1 mg/ml | |
| Western Blot 1:1000-1:10000, ELISA 1:5000-1:50000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:50-1:200, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating, Knockout Validated, Super-Resolution Microscopy | |
| P02462 | |
| COL4A1 | |
| Collagen IV from human and bovine placenta (Uniprot: P02462) | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Some class-specific anti-collagens may be specific for three-dimensional epitopes which may result in diminished reactivity with denatured collagen or formalin-fixed, paraffin embedded tissues. This antibody reacts with most mammalian Collagen IV and has negligible cross-reactivity with Type I, II, III, V or VI collagens. Non-specific cross-reaction of anti-collagen antibodies with other human serum proteins or non-collagen extracellular matrix proteins is negligible. | |
| Store at 4°C short term. For extended storage, add an equal volume of glycerol, aliquot and store at -20°C or below. Avoid repeated freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction