missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen VI alpha 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 12 publications
Brand: Novus Biologicals NB120-6588SS
This item is not returnable.
View return policy
Description
Collagen VI alpha 1 Polyclonal specifically detects Collagen VI alpha 1 in Human, Mouse, Rat, Bovine, Feline samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, PCR.
Specifications
| Collagen VI | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| Collagen 6, collagen alpha-1(VI) chain, collagen VI, alpha-1 polypeptide, collagen, type VI, alpha 1, Collagen-6, human mRNA for collagen VI alpha-1 C-terminal globular domain10alpha 1 (VI) chain (61 AA), OPLL | |
| Rabbit | |
| 108.53 kDa | |
| 0.025 mg | |
| Cytoskeleton Markers, Extracellular Matrix | |
| 1291 | |
| Human, Mouse, Rat, Bovine, Feline, Mammalia | |
| IgG |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 1 mg/ml | |
| Western Blot 1:1000-1:10000, Flow Cytometry, ELISA 1:5000-1:50000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:50-1:200 | |
| P12109 | |
| COL6A1 | |
| Collagen VI alpha 1 from human and bovine placenta (Uniprot: P12109) | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Some class-specific anti-collagens may be specific for three-dimensional epitopes which may result in diminished reactivity with denatured collagen or formalin-fixed, paraffin embedded tissues. This antibody reacts with most mammalian Collagen VI alpha 1 and has negligible cross-reactivity with Type I, II, III, IV or V collagens. Non-specific cross-reaction of anti-collagen antibodies with other human serum proteins or non-collagen extracellular matrix proteins is negligible. | |
| Store at 4°C short term. For extended storage, add an equal volume of glycerol, aliquot and store at -20°C or below. Avoid repeated freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction