missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Collectrin/TMEM27 Antibody [Alexa Fluor™ 532], Novus Biologicals™

Product Code. 30042869
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30042869 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30042869 Supplier Novus Biologicals™ Supplier No. NBP330745AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Collectrin/TMEM27 Polyclonal antibody specifically detects Collectrin/TMEM27 in Human samples. It is validated for ELISA,Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Collectrin/TMEM27
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50 mM Sodium Borate
Gene Alias collectrin, kidney-specific membrane protein, NX-17, transmembrane protein 27NX170610008J07Rik
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human Collectrin/TMEM27 (RefSeq#: NP_065716.1; Met1-Pro141)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 57393
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.