Learn More
Novus Biologicals™ Colon Matched Tumor and Normal Tissue Lysate
Product Code. 18169898 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
2 Vials
Unit Size:
Pack of 2
Product Code. Quantity Unit Size
18169898 2 Vials Pack of 2
1 options
Product Code. 18169898 Supplier Novus Biologicals™ Supplier No. NBP2470782VIAL
  • ONLINE EXCLUSIVE PRICE
    685.00 €
    647.85 € / Pack of 2
    Save 37.15 €
    5% Off
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Two matched 1 mg/mL vials (100 μg/vial) of human colon tissue lysate, one containing normal tissue and the other tumor tissue, for use in research. Lysates are guaranteed for 6 months from date of receipt.
The Colon Matched Tumor and Normal Tissue Lysate is derived from human.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 2 Vials
Content And Storage Store at -80C. Avoid freeze-thaw cycles.