missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COLQ Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92021-0.1ml
This item is not returnable.
View return policy
Description
COLQ Polyclonal antibody specifically detects COLQ in Mouse, Rat samples. It is validated for Western Blot
Specifications
| COLQ | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| acetylcholinesterase collagenic tail peptide, AChE Q subunit, collagenic tail of endplate acetylcholinesterase, collagen-like tail subunit (single strand of homotrimer) of asymmetricacetylcholinesterase, EADAcetylcholinesterase-associated collagen, FLJ55041, single strand of homotrimeric collagen-like tail subunit of asymmetricacetylcholinesterase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 276-455 of human COLQ (NP_005668.2). MGPKGERGFPGPPGRCLCGPTMNVNNPSYGESVYGPSSPRVPVIFVVNNQEELERLNTQNAIAFRRDQRSLYFKDSLGWLPIQLTPFYPVDYTADQHGTCGDGLLQPGEECDDGNSDVGDDCIRCHRAYCGDGHRHEGVEDCDGSDFGYLTCETYLPGSYGDLQCTQYCYIDSTPCRYFT | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 8292 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction