missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Columbia Agar with Horse Blood

Product Code. 12971130
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12971130 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12971130 Supplier Thermo Scientific™ Supplier No. PB0122A
  • ONLINE EXCLUSIVE PRICE
    19.95 €
    17.06 € / Pack of 10
    Save 2.89 €
    14% Off
View all promotions
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cultivate and determine haemolytic reactions of fastidious organisms with this multipurpose microbiological medium containing 5% horse blood.

Cultivate, isolate and identify a broad range of microorganisms, using Thermo Scientific™ Columbia Agar with Horse Blood. Rapid production of large colonies, clearly defined zones of haemolysis and good colonial differentiation is promoted by the addition of defibrinated horse blood.

Traditionally, blood agar bases have been either casein hydrolysate or meat infusion media. The advantage of the first lies in the rapid production of large colonies, and of the second in clearly defined zones of haemolysis and good colonial differentiation. Columbia Agar Base combines the virtues of both these types of media to give an improved all-round performance, showing versatility and superior performance in several applications.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Color Red, opaque
Content And Storage 2°C to 10°C
Description Columbia Agar with Horse Blood
Format 90mm Monoplate
Manufacturing Origin United Kingdom
Product Type Agar Plate
Fill Weight 19.5 g ± 1.0 g
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.