missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Columbia Blood Agar with Neomycin

Product Code. 12982598
Change view
Click to view available options
Description:
Columbia Blood Agar with Neomycin
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
12982598 Columbia Blood Agar with Neomycin Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12982598 Supplier Thermo Scientific™ Supplier No. PB0219A
  • ONLINE EXCLUSIVE PRICE
    20.10 €
    19.58 € / Pack of 10
    Save 0.52 €
    2% Off
View all promotions
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Cultivate and isolate a wide variety of fastidious microorganisms with this multi purpose, ready-to-use medium.

Cultivate and isolate a wide variety of fastidious microorganisms with Thermo Scientific™ Oxoid™ Columbia Blood Agar with Neomycin. Columbia Agar is an ideal base medium for the addition of blood especially in selective formulations when supplemented with antimicrobial agents. The addition of defibrinated horse blood, in this case, facilitates cultivation of fastidious microorganisms. Neomycin creates a selectively anaerobic environment for the cultivation of anaerobes such as clostridia species.

Traditionally, blood agar bases have been either casein hydrolysate or meat infusion media. The advantage of the first lies in the rapid production of large colonies and of the second in clearly defined zones of hemolysis and good colonial differentiation. Oxoid™ Columbia Blood Agar with Neomycin combines the virtues of both of these types of media to give an improved all-round performance, showing versatility and superior performance in several applications.1

  • Facilitates growth of large colonies of fastidious microorganisms with clearly defined hemolytic reactions and colonial morphology
  • Select for anaerobic bacteria because of the addition of the selective agent Neomycin
  • Ready-to-use convenience of prepared media

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Red
Content And Storage 2°C to 10°C
Description Columbia Blood Agar with Neomycin
Format 90mm Monoplate
Manufacturing Origin United Kingdom
Product Type Agar Plate
Fill Weight 19.5g±1.0.g
Packaging Type 10 x 90mm plates, film wrapped
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.