missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COM1/p8 Antibody [Unconjugated], Novus Biologicals™
Rabbit Polyclonal Antibody
422.00 € - 607.00 €
Specifications
| Antigen | COM1/p8 |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL |
| Applications | Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30279463
|
Novus Biologicals
NBP3-43682-25ul |
25 μL |
422.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30279396
|
Novus Biologicals
NBP3-43682-100ul |
100 μL |
607.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
COM1/p8 Polyclonal antibody specifically detects COM1/p8 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| COM1/p8 | |
| Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| PBS, pH 7.2, 40% glycerol | |
| 26471 | |
| IgG | |
| Affinity purified |
| Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Candidate of metastasis 1, COM1P8, nuclear protein 1, nuclear protein, transcriptional regulator, 1, p8, Protein p8 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: TFPPATSAPQQPPGPEDEDSSLDESDLYSLAHSYLGGGGRKGRTKREAAANTNRPSPGGHERKLVTKLQNSERKK | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title