Learn More
CombiCheck™ 18701 100 Multi-Parameter Standard Solution, WTW™
Product Code. 15715795
Change view
Click to view available options
:
0.40 mg/L Al; 2.00 mg/L Ni; 2.00 mg/L Pb; 0.75 mg/L Zn
Product Code.
15715795 0.40 mg/L Al; 2.00 mg/L Ni; 2.00 mg/L Pb; 0.75 mg/L Zn
1 options
Product Code. 15715795 Supplier WTW Supplier No. 252502
Edge
WTW™ CombiCheck™ Solutions are ready-to-use multi-parameter standards. Every package contains a standard solution and an addition solution. Both solutions can be used directly without dilution, for quality assurance purposes.
  • Ready-to-use multi-parameter photometric standards
  • The standard solution ensures the accuracy of the results from the entire system
  • The addition solution allows you to check sample-dependent influences
  • Up to 280 determinations are possible using the addition solution
TRUSTED_SUSTAINABILITY
Specifications
Concentration or Composition Notes 0.40 mg/L Al; 2.00 mg/L Ni; 2.00 mg/L Pb; 0.75 mg/L Zn