missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Common beta Chain Antibody (BION-1) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
251.00 € - 655.00 €
Specifications
| Antigen | Common beta Chain |
|---|---|
| Clone | BION-1 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Functional Assay, Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18607046
|
Novus Biologicals
NBP3-09024-0.025mg |
0.025 mg |
251.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18618437
|
Novus Biologicals
NBP3-09024-0.2mg |
0.2 mg |
655.00 €
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Common beta Chain Monoclonal antibody specifically detects Common beta Chain in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Functional Assay, NeutralizationSpecifications
| Common beta Chain | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| 1439 | |
| IgG1 κ | |
| Protein A purified |
| BION-1 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Functional Assay, Neutralization | |
| Unconjugated | |
| Mouse | |
| Human | |
| beta, low-affinity, CD131, CD131 antigen, CDw131, colony stimulating factor 2 receptor, beta, low-affinity(granulocyte-macrophage), cytokine receptor common subunit beta, GM-CSF/IL-3/IL-5 receptor common beta subunit, GM-CSF/IL-3/IL-5 receptor common beta-chain, IL3RB, interleukin 3 receptor/granulocyte-macrophage colony stimulating factor 3receptor, beta (high affinity) | |
| BALB/c mice were immunized intraperitonally with COS cells transfected DQP (Domain 4 of the common beta-chain) expression constructs. The immunizations were repeated 4 times at 2-week intervals. Four weeks after the final immunization, a mouse was boosted with COS transfectants intravenously. Three days later, splenocytes were harvested and fused with NS-1 myeloma cells. Hybridoma supernatants were screened on CHO DQP cells by flow cytometry, with untransfected CHO cells as a control. All antibodies were from single hybridoma clones as selected by a limiting dilution method. MoAbs were purified from ascites fluid or hybridoma supernatant by a protein A sepharose column. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title