missing translation for 'onlineSavingsMsg'
Learn More

Common Elements Mix 3: Ag, Al, B, Ca, Co, Cr, Cu, Fe, K, Li, Mg, Mn, Mo, Na, Ni, P, Pb, Si, Sn, Ti, V, Zn @ 100 μg/mL in 5% HNO3, tr. HF, VHG™

Product Code. 17957929
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17957929 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 17957929 Supplier VHG Labs Supplier No. VHGSM70C500
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Common Elements Mix 3: Ag, Al, B, Ca, Co, Cr, Cu, Fe, K, Li, Mg, Mn, Mo, Na, Ni, P, Pb, Si, Sn, Ti, V, Zn @ 100 μg/mL in 5% HNO3, tr. HF
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Boron/Calcium/Chromium/Cobalt/Copper/Iron/Lead/Lithium/Magnesium/Manganese/Molybdenum/Nickel/Phosphorus/Potassium/Silicon/Silver/Sodium/Tin/Titanium/Vanadium/Zinc/Aluminium
CAS 7440-42-8, 7440-70-2, 7440-47-3, 7440-48-4, 7440-50-8, 7439-89-6
Methods ICP/ICPMS
Analyte or Component Names Boron/Calcium/Chromium/Cobalt/Copper/Iron/Lead/Lithium/Magnesium/Manganese/Molybdenum/Nickel/Phosphorus/Potassium/Silicon/Silver/Sodium/Tin/Titanium/Vanadium/Zinc/Aluminium
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix 5% HNO3, tr. HF
Quantity 500 mL
Concentration or Composition (by Analyte or Components) Boron 100 μg/mL/Calcium 100 μg/mL/Chromium 100 μg/mL/Cobalt 100 μg/mL/Copper 100 μg/mL/Iron 100 μg/mL/Lead 100 μg/mL/Lithium 100 μg/mL/Magnesium 100 μg/mL/Manganese 100 μg/mL/Molybdenum 100 μg/mL/Nickel 100 μg/mL/Phosphorus 100 μg/mL/Potassium 100 μg/mL/Silicon 100 μg/mL/Silver 100 μg/mL/Sodium 100 μg/mL/Tin 100 μg/mL/Titanium 100 μg/mL/Vanadium 100 μg/mL/Zinc 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.