missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ COMP Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA596435

Product Code. 16569271

  • 520.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: GQGQSPLGSD LGPQMLRELQ ETNAALQDVR ELLRQQVREI TFLKNTVMEC DACGMQQSVR TGLPSVRPLL HCAPGFCFPG VACIQTESGA RCGPCPAGFT GNGSHCTDVN ECNAHPCFPR VRCINTSPGF RCEACPPGYS G; Positive Samples: SW620, U-251MG, HepG2, MCF7, Mouse skeletal muscle; Cellular Location: extracellular matrix, extracellular space.

COMP is a noncollagenous extracellular matrix protein. It consists of five identical glycoprotein subunits, each with EGF like and calcium binding (thrombospondin like) domains. Oligomerization results from formation of a five stranded coiled coil and disulfides. Binding to other ECM proteins such as collagen appears to depend on divalent cations. Mutations can cause the osteochondrodysplasias pseudochondroplasia (PSACH) and multiple epiphyseal dysplasia (MED).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

COMP
Polyclonal
Unconjugated
COMP
Cartilage oligomeric matrix protein; cartilage oligomeric matrix protein (pseudoachondroplasia, epiphyseal dysplasia 1, multiple); cartilage oligomeric matrix protein(pseudoachondroplasia, epiphyseal dysplasia 1, multiple); Comp; EDM1; EPD1; MED; MGC131819; MGC149768; PSACH; pseudoachondroplasia (epiphyseal dysplasia 1, multiple); putative cartilage oligomeric matrix protein; THBS5; Thrombospondin5; Thrombospondin-5; TSP 5; TSP5
Rabbit
Affinity Chromatography
RUO
12845, 1311, 25304
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot
2.56 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
P35444, P49747, Q9R0G6
COMP
Recombinant protein (or fragment).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.