missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C6 Antibody (12), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-05829-50ul
This item is not returnable.
View return policy
Description
Complement C6 Monoclonal antibody specifically detects Complement C6 in Human samples. It is validated for ELISA
Specifications
| Complement C6 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 12 | |
| ELISA 1:1000-1:2000 | |
| complement component 6, complement component C6 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Complement C6 (Accession#: AAA59668.1; Met1-Ala934). | |
| 50 μg | |
| Immunology, Innate Immunity | |
| 729 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction