missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Factor H-related 3/CFHR3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92615-0.1ml
This item is not returnable.
View return policy
Description
Complement Factor H-related 3/CFHR3 Polyclonal antibody specifically detects Complement Factor H-related 3/CFHR3 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Complement Factor H-related 3/CFHR3 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| CFHL3;Complement factor H-related protein 3;DOWN16;FHR3;FHR-3;HLF4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 80-200 of human CFHR3 (NP_066303.2). AVPCLRKCYFPYLENGYNQNYGRKFVQGNSTEVACHPGYGLPKAQTTVTCTEKGWSPTPRCIRVRTCSKSDIEIENGFISESSSIYILNKEIQYKCKPGYATADGNSSGSITCLQNGWSAQ | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10878 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction