missing translation for 'onlineSavingsMsg'
Learn More

Arsenic Concentrate, 10ppm, EP Grade, Reagecon™
BIOPHARMA

Product Code. 11752987 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11752987 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11752987 Supplier Reagecon Supplier No. 5000500C
  • ONLINE EXCLUSIVE PRICE
    110.00 €
    108.86 € / 100mL
    Save 1.14 €
    1% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Concentrated to make arsenic standard solution. Arsenic Concentrate, 10ppm, EP Grade, Reagecon™ is manufactured according to European Pharmacopoeia Chapter 4(4.1.2) for use in arsenic limit tests.

  • Manufactured in an ISO 9001-compliant facility according to Eu. Ph. Chapter 4(4.1.2) guidelines
  • Produced in accordance with ASTM, APHA, ACS, EP and USP methods
  • Provided with a certificate of analysis and a label that shows the lot and expiration date
  • Ready to use solution eliminates the time and labor of preparing precise standard solutions

Specifications

CAS 7440-38-2
Molecular Formula As
MDL Number MFCD00085309
Concentration 10ppm As
Synonym arsenicals, arsenic, inorganic, arsenic-75, arsen, gray, grey, black, metallic, compounds, colloidal
InChI Key RQNWIZPPADIBDY-UHFFFAOYSA-N
SMILES [As]
IUPAC Name arsenic
Molecular Weight (g/mol) 74.92
PubChem CID 5359596
ChEBI CHEBI:27563
Formula Weight 74.92g/mol
Grade Eu. Ph. Chapter 4(4.1.2)
Odor Odorless
Methods Arsenic Limit Tests
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Arsenic
Show More Show Less
compliance-icons
Product Identifier
  • CONCENTRATE TO MAKE (5000500)
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1A
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: diarsenic trioxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.