missing translation for 'onlineSavingsMsg'
Learn More

Lead Concentrate, 100ppm, EP Grade, Reagecon™
BIOPHARMA

Product Code. 11782048 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. Quantity unitSize
11782048 100 mL 100mL
1 options
This item is not returnable. View return policy

Product Code. 11782048

Brand: Reagecon 5001701C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Concentrated to make lead standard solution. Lead Concentrate, 100ppm, EP Grade, Reagecon™ is a lead nitrate solution with 100ppm Pb. It is manufactured according to European Pharmacopoeia Chapter 4(4.1.2) for use in lead determination and lead limit tests.

  • Manufactured in an ISO 9001-compliant facility according to Eu. Ph. Chapter 4(4.1.2) guidelines
  • Produced in accordance with ASTM, APHA, ACS, EP and USP methods
  • Provided with a certificate of analysis and a label that shows the lot and expiration date
  • Ready to use solution eliminates the time and labor of preparing precise standard solutions

Specifications

Color Colorless
CAS 10099-74-8
Molecular Formula N2O6Pb
MDL Number MFCD00011153
Concentration 1000ppm Pb
Synonym lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt
InChI Key RLJMLMKIBZAXJO-UHFFFAOYSA-N
SMILES [Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
IUPAC Name lead(2+);dinitrate
Molecular Weight (g/mol) 331.20
PubChem CID 24924
ChEBI CHEBI:37187
Formula Weight 331.2g/mol
Grade Eu. Ph. Chapter 4(4.1.2)
Odor Odorless
Methods Lead Limit Tests
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Lead Nitrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.