missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Solution Standard, Hanna Instruments

Product Code. 11574317
Change view
Click to view available options
Quantity:
460 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11574317 460 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11574317 Supplier Hanna Instruments Supplier No. HI70300L
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A storage solution prepared with reagent grade chemicals that can be used to ensure optimum performance of your pH and ORP electrodes.

  • Storage solution prepared with reagent grade chemicals that can be used to ensure optimum performance of your pH and ORP electrodes
  • Special formulation to minimize microbial growth and osmotic/diffusion effects between the solution and inner reference electrolyte
  • Manufactured specifically for pH and ORP electrodes
  • Clearly marked expiration date and lot number

Specifications

Chemical Name or Material Conductivity Solution Standard
Quantity 460 mL
compliance-icons
Product Identifier
  • ORP Solution for Platinum and Gold Electrodes
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Sulphuric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.