missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Conductivity Standard 10 μS/cm at 25 °C in 30% n-propanol
Shop All Fisher Chemical ProductsDescription
- Calibration: Ensures conductivity meters give accurate readings.
- Quality control: Used in water testing, environmental monitoring, and industrial processes.
- Traceability: Certified standards provide documented accuracy and uncertainty.
- Temperature compensation: Standards are often referenced at 25°C, since conductivity varies strongly with temperature.
- Certified Reference Material (CRM) according to ISO 17025 and ISO 17034
- Each CRM comes with a certificate of analysis that includes uncertainty calculations.
- This documentation is critical for audits, method validation, and proving the reliability of your data.
Specifications
Specifications
| Quantity | 500 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?