missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard Solutions, Hanna Instruments™

Product Code. 10309960
Change view
Click to view available options
Quantity:
20 mL
450 mL
500 mL
Conductivity:
12,880 μS/cm
1413 μS/cm
84 μS/cm
Unit Size:
450mL
Each
Pack of 25
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Conductivity unitSize
10309960 20 mL 12,880 μS/cm Pack of 25
10441141 20 mL 1413 μS/cm Pack of 25
12776955 450 mL 1413 μS/cm 450mL
12786955 450 mL 84 μS/cm 450mL
11514327 500 mL 1413 μS/cm Each
11574327 500 mL 84 μS/cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10309960 Supplier Hanna Instruments Supplier No. HI70030P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A premium quality conductivity standard that is prepared against a NIST traceable potassium chloride solution.

  • Premium quality 12,880 μS/cm conductivity solution that is NIST traceable and contained in portable single-use sachets. 
  • Temperature chart of the actual μS/cm value at various temperatures printed on each bottle
  • Premium quality 12,880 μS/cm conductivity solution standardized with a conductivity meter that has been calibrated with potassium chloride NIST 2202 SRM (Standard Reference Material) 
  • Deionized water for analytical use in accordance with ISO 3696/BS3978
  • Ideal for users that do not calibrate frequently
  • Ideal for field use due to portability

Specifications

Conductivity 12,880 μS/cm
Type Calibration Standard
Industry Type Laboratory Use, Industrial Use
Quantity 20 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.