missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard, NIST Traceable, 200,000μS/cm, Reagecon™
BIOPHARMA

Product Code. 11576723 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Product Code. Quantity unitSize
11576723 500 mL 500mL
1 options
This item is not returnable. View return policy

Product Code. 11576723

Brand: Reagecon CSKC200M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Accredited to ISO 17025. Reagecon™ Conductivity Standard, NIST Traceable, 200,000μS/cm is an aqueous-based, non-hazardous standard with a high degree of accuracy, low temperature variation and guaranteed stability through the expiration date, even after opening the bottle.

This conductivity standard is aqueous based, thereby eliminating any errors attributable to matrix mismatch. Aqueous standards are non-hazardous for easy storage, handling and disposal. Each bottle is supplied with a comprehensive Certificate of Analysis and has a table of conductivity variation with temperature printed on the label.

  • Convenience and lower costs—Reduced health, safety and environmental requirements and paperwork
  • Very low temperature coefficient of variation—Reduces measurement errors and enables non-temperature controlled conditions or field work
  • Guaranteed stability through expiry date—Eliminates the need to open a fresh bottle of standard every time it is used
  • High accuracy (±1% at 25°C) —Enables it to be used as a calibrator or control, in accordance with the most exacting requirements (e.g. USP)

Specifications

Certificate of Analysis Yes
Conductivity 200,000μS/cm (±1% at 25°C)
Physical Form Liquid
For Use With (Equipment) Conductivity Meter
Solvent or Matrix Water
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.