missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This conductivity standard is aqueous based, thereby eliminating any errors attributable to matrix mismatch. Aqueous standards are non-hazardous for easy storage, handling and disposal. Each bottle is supplied with a comprehensive Certificate of Analysis and has a table of conductivity variation with temperature printed on the label.
- Convenience and lower costs—Reduced health, safety and environmental requirements and paperwork
- Very low temperature coefficient of variation—Reduces measurement errors and enables non-temperature controlled conditions or field work
- Guaranteed stability through expiry date—Eliminates the need to open a fresh bottle of standard every time it is used
- High accuracy (±1% at 25°C) —Enables it to be used as a calibrator or control, in accordance with the most exacting requirements (e.g. USP)
Specifications
Specifications
| Certificate of Analysis | Yes |
| Conductivity | 300,000μS/cm (±1% at 25°C) |
| Physical Form | Liquid |
| For Use With (Equipment) | Conductivity Meter |
| Solvent or Matrix | Water |
| Quantity | 500 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
