missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard, NIST Traceable, 5,000μS/cm, Reagecon™
Click to view available options
Quantity:
500 mL
Description
This conductivity standard is aqueous based, thereby eliminating any errors attributable to matrix mismatch. Aqueous standards are non-hazardous for easy storage, handling and disposal. Each bottle is supplied with a comprehensive Certificate of Analysis and has a table of conductivity variation with temperature printed on the label.
- Convenience and lower costs—Reduced health, safety and environmental requirements and paperwork
- Very low temperature coefficient of variation—Reduces measurement errors and enables non-temperature controlled conditions or field work
- Guaranteed stability through expiry date—Eliminates the need to open a fresh bottle of standard every time it is used
- High accuracy (±1% at 25°C) —Enables it to be used as a calibrator or control, in accordance with the most exacting requirements (e.g. USP)
Specifications
Quantity | 500 mL |
Certificate of Analysis | Yes |
For Use With (Equipment) | Conductivity Meter |
Solvent or Matrix | Aqueous |
Conductivity | 5,000μS/cm (±1% at 25°C) |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'