missing translation for 'onlineSavingsMsg'
Learn More

Coniferyl alcohol, 98%

Product Code. 11421487
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11421487 0.25 g 0.25g
11401487 1 g 1g
11411487 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11421487 Supplier Thermo Scientific Alfa Aesar Supplier No. B24949.01
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 458-35-5 | C10H12O3 | 180.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 458-35-5
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00002922
InChI Key JMFRWRFFLBVWSI-NSCUHMNNSA-N
Synonym coniferyl alcohol, coniferol, coniferylalcohol, 4-3-hydroxyprop-1-en-1-yl-2-methoxyphenol, gamma-hydroxyisoeugenol, trans-coniferyl alcohol, e-coniferyl alcohol, 4-3-hydroxy-1-propenyl-2-methoxyphenol, 3-4-hydroxy-3-methoxyphenyl-2-propen-1-ol, 4-1e-3-hydroxyprop-1-en-1-yl-2-methoxyphenol
PubChem CID 1549095
ChEBI CHEBI:17745
SMILES COC1=CC(\C=C\CO)=CC=C1O

Specifications

Melting Point 75°C to 76°C
Boiling Point 163°C to 165°C (3 mmHg)
Quantity 0.25 g
Beilstein 2048961
Sensitivity Air and light sensitive
Merck Index 14,2504
IUPAC Name 4-[(1E)-3-hydroxyprop-1-en-1-yl]-2-methoxyphenol
Formula Weight 180.2
Percent Purity 98%
Chemical Name or Material Coniferyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.