missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contactin-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98360-50ul
This item is not returnable.
View return policy
Description
Contactin-1 Polyclonal antibody specifically detects Contactin-1 in Mouse samples. It is validated for ELISA
Specifications
| Contactin-1 | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| contactin 1, contactin-1, F3, Glycoprotein gp135, GP135, Neural cell surface protein F3 | |
| Produced in rabbits immunized with purified, recombinant Mouse Contactin-1 (Accession#: NP_031753.1; Met1-Leu1000) | |
| 50 μg | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 1272 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction