missing translation for 'onlineSavingsMsg'
Learn More

Coomassie Brilliant Blue G-250, MP Biomedicals™

Product Code. 11465951 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11465951 25 g 25g
11455951 5 g 5g
11441642 10 g 10g
11475951 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11465951 Supplier MP Biomedicals Supplier No. 0219034325

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Useful dye for SDS gels as it readily stains proteins with minimal background color

  • It is a protein stain in electrophoresis.
  • It has been used in the Bradford dye-binding protein assay.
  • A mechanism for dye binding to protein has been proposed, based on measuring Coomassie Brilliant Blue G-250 (CBBG) absorbance spectra during titration of the dye reagent in the absence of protein and its response to different polyamino acids.
  • Brilliant Blue G has been used for determining critical micelle concentration of detergents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6104-58-1
Molecular Formula C47H48N3NaO7S2
PubChem CID 6328534
IUPAC Name sodium;3-[[4-[(Z)-[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]-2-methylcyclohexa-2,5-dien-1-ylidene]methyl]-N-ethyl-3-methylanilino]methyl]benzenesulfonate
SMILES CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC(=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3C)C5=CC=C(C=C5)NC6=CC=C(C=C6)OCC)C.[Na+]

Specifications

Synonym Acid Blue 90, Brilliant Blue G
InChI Key RWVGQQGBQSJDQV-UHFFFAOYSA-M
Molecular Weight (g/mol) 854.025
Formula Weight 854
Grade Electrophoresis
Color Blue to Purple
Quantity 25 g
Physical Form Powder
Chemical Name or Material Coomassie Brilliant Blue G-250, Protein stain in electrophoresis.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.