missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COP1 Antibody (1E4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00064326-M01
This item is not returnable.
View return policy
Description
COP1 Monoclonal antibody specifically detects COP1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA
Specifications
| COP1 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| Constitutive photomorphogenesis protein 1 homolog, constitutive photomorphogenic protein (COP1), COP1RNF200FLJ10416, E3 ubiquitin-protein ligase RFWD2, EC 6.3.2.-, hCOP1, putative ubiquitin ligase COP1, ring finger and WD repeat domain 2, RING finger and WD repeat domain protein 2, RING finger protein 200 | |
| RFWD2 (NP_071902, 632 a.a. ∽ 731 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. CLRSFKGHINEKNFVGLASNGDYIACGSENNSLYLYYKGLSKTLLTFKFDTVKSVLDKDRKEDDTNEFVSAVCWRALPDGESNVLIAANSQGTIKVLELV | |
| 0.1 mg | |
| p53 Pathway, Zinc Finger | |
| 64326 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Sandwich ELISA | |
| 1E4 | |
| Western Blot 1:500, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 1:10-1:500, Sandwich ELISA | |
| NP_071902 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction