missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COPG2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00 € - 463.00 €
Specifications
| Antigen | COPG2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/ Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18695502
|
Novus Biologicals
NBP2-92868-0.02ml |
0.02 mL |
190.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18664852
|
Novus Biologicals
NBP2-92868-0.1ml |
0.1 mL |
463.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
COPG2 Polyclonal antibody specifically detects COPG2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| COPG2 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Golgi Apparatus Markers, Membrane Trafficking and Chaperones | |
| PBS with 50% glycerol, pH7.3. | |
| 26958 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/ Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 2-COP, coat protein, nonclathrin, gamma-2-cop, coatomer protein complex, subunit gamma 2, coatomer subunit gamma-2, DKFZp761N09121, FLJ11781, Gamma-2-coat protein, gamma-2-COP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 542-871 of human COPG2 (NP_036265.3). LNATYIFNGLTVSVPGMEKALHQYTLEPSEKPFDMKSIPLAMAPVFEQKAEITLVATKPEKLAPSRQDIFQEQLAAIPEFLNIGPLFKSSEPVQLTEAETEYFVRCIKHMFTNHIVFQFDCTNTLNDQLLEKVTVQMEPSDSYEVLSCIPAPSLPYNQPGICYTLVRLPDDDPTAVAGSFSCTMKFTVRDCDPNTGVPDEDGYDDEYVLEDLEVTVSDHIQKVLKPNFAAAWEEVGDTFEKEETFALSSTKTLEEAVNNIITFLGMQPCERSDKVPENKNSHSLYLAGIFRGGYDLLVRSRLALADGVTMQVTVRSKERTPVDVILASVG | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title