missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The coatomer is a cytosolic protein complex that binds to dilysine motifs and reversibly associates with Golgi non-clathrin-coated vesicles, which further mediate biosynthetic protein transport from the ER, via the Golgi up to the trans Golgi network.
Specifications
Specifications
| Antigen | COPG2 |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.37 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | COPG2 |
| Gene Accession No. | Q9QXK3, Q9UBF2 |
| Gene Alias | 2 COP, Coatomer subunit gamma 2, COPG2, Gamma 2 coat protein, Gamma 2 COP |
| Gene Symbols | COPG2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?