missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) bromide-dimethyl sulfide complex

Product Code. 11343537
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343537 10 g 10g
11333537 2 g 2g
11323537 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11343537 Supplier Thermo Scientific Alfa Aesar Supplier No. 019368.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 54678-23-8 | C2H6BrCuS | 205.58 g/mol

Copper(I) bromide-dimethyl sulfide complex is used as catalyst for adding Grignard reagents to fullerenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54678-23-8
Molecular Formula C2H6BrCuS
Molecular Weight (g/mol) 205.58
MDL Number MFCD00043295
InChI Key PMHQVHHXPFUNSP-UHFFFAOYSA-M
Synonym copper i bromide-dimethyl sulfide, bromo thiobis methane copper, copper i bromide dimethyl sulfide complex, bromocopper; methylsulfanylmethane, cubr.me2s, ksc493e2t, copper i bromide dimethylsulfide, copper i bromide-dimethylsulfide, bromo dimethylsulfide copper i
PubChem CID 2724234
SMILES [Cu]Br.CSC

Specifications

Melting Point 124°C to 129°C (decomposition)
Color Brown or Green
Physical Form Crystalline
Quantity 10 g
UN Number UN3335
Solubility Information Soluble in water. Reacts with water and decomposes.
IUPAC Name (methylsulfanyl)methane; bromocopper
Formula Weight 205.58
Sensitivity Air and moisture sensitive
Chemical Name or Material Copper(I) bromide-dimethyl sulfide complex

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.