missing translation for 'onlineSavingsMsg'
Learn More

Copper standard solution, for AAS, 1 mg/ml Cu in 2 to 5% HNO3

Product Code. 10419900 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10419900 500 mL 500mL
10666022 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10419900 Supplier Thermo Scientific Acros Supplier No. 195935000
  • ONLINE EXCLUSIVE PRICE
    64.40 €
    60.10 € / 500mL
    Save 4.30 €
    6% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7440-50-8 | Cu | 63.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Copper standard solution
Name Note 1 mg/ml Cu in 2% HNO3
Packaging Plastic bottle
CAS 7440-50-8, 7697-37-2, 7732-18-5
Density 8.92
Physical Form Solution
Molecular Formula Cu
Concentration 997 to 1003 μg/mL of Cu(2+) in 2% HNO3 (20°C)
MDL Number MFCD00010965
Color Blue
Fieser 16,95; 16,181
Solubility Soluble in water
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES [Cu]
IUPAC Name copper
Molecular Weight (g/mol) 63.55
PubChem CID 23978
Quantity 500 mL
ChEBI CHEBI:30052
Formula Weight 63.55
Grade AAS
Show More Show Less
compliance-icons
Product Identifier
  • Copper atomic absorption standard solution
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.