missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COQ7 Antibody (06), Novus Biologicals™
Mouse Monoclonal Antibody
220.00 € - 326.00 €
Specifications
| Antigen | COQ7 |
|---|---|
| Clone | 06 |
| Dilution | Sandwich ELISA Capture 1:250-1:2000 |
| Applications | Sandwich ELISA Capture |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18616977
|
Novus Biologicals
NBP3-05806-100ul |
100 μg |
326.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18659747
|
Novus Biologicals
NBP3-05806-50ul |
50 μg |
220.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
COQ7 Monoclonal antibody specifically detects COQ7 in Human samples. It is validated for Sandwich ELISA CaptureSpecifications
| COQ7 | |
| Sandwich ELISA Capture 1:250-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CAT5, CLK1, CLK-1, Coenzyme Q biosynthesis protein 7 homolog, coenzyme Q, 7 (rat, yeast) homolog, coenzyme Q7 homolog, ubiquinone (yeast), COQ7 coenzyme Q, 7 homolog ubiquinone, placental protein KG-20, Timing protein clk-1 homolog, ubiquinone biosynthesis protein COQ7 homolog | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human COQ7 (Uniprot#: Q99807-1; Ser37-Leu217). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 06 | |
| Sandwich ELISA Capture | |
| Unconjugated | |
| Mouse | |
| Stem Cell Markers | |
| 0.2 um filtered solution in PBS | |
| 10229 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title