missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COQ9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99187-100ul
This item is not returnable.
View return policy
Description
COQ9 Polyclonal antibody specifically detects COQ9 in Human samples. It is validated for Western Blot
Specifications
| COQ9 | |
| Polyclonal | |
| Western Blot 1:500-1:1000 | |
| C16orf49DKFZp434K046, chromosome 16 open reading frame 49, coenzyme Q9 homolog (S. cerevisiae), coenzyme Q9 homolog (yeast), DKFZP434K046, ubiquinone biosynthesis protein COQ9, mitochondrial | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human COQ9. | |
| 100 μg | |
| Cancer, Endocrinology, Signal Transduction | |
| 57017 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction