missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 405 Anti-Mouse CD4 (GK1.5), Proteintech
Rat Monoclonal Antibody
Brand: Proteintech CL405-65104-100UG
This item is not returnable.
View return policy
Description
The CD4 antibody from Proteintech is a rat monoclonal antibody generated with mouse ctl clone v4 of mouse CD4. This antibody recognizes mouse antigen. The CD4 antibody has been validated for the following applications: FC analysis. This antibody is coralite™ plus 405.Specifications
| CD4 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Rat | |
| Affinity purification | |
| RUO | |
| 12504 | |
| Store at 2-8°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Flow Cytometry | |
| GK1.5 | |
| CoraLite Plus 405 | |
| Mouse CTL clone V4 | |
| 100 μg | |
| Primary | |
| Mouse | |
| Flow Antibody | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction