missing translation for 'onlineSavingsMsg'
Learn More

Corilagin, TRC

Product Code. 30349499
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30349499 5 mg 5mg
30359520 1 mg 1mg
30364596 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30349499 Supplier Toronto Research Chemicals Supplier No. TRCC6954005MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23094-69-1
Molecular Formula C27 H22 O18
Molecular Weight (g/mol) 634.45
IUPAC Name [(1S,19R,21S,22R,23R)-6,7,8,11,12,13,22,23-octahydroxy-3,16-dioxo-2,17,20-trioxatetracyclo[17.3.1.04,9.010,15]tricosa-4,6,8,10,12,14-hexaen-21-yl] 3,4,5-trihydroxybenzoate
SMILES O[C@@H]1[C@H]2COC(=O)c3cc(O)c(O)c(O)c3c4c(O)c(O)c(O)cc4C(=O)O[C@@H]1[C@@H](O)[C@H](OC(=O)c5cc(O)c(O)c(O)c5)O2

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C27H22O18/c28-9-1-6(2-10(29)16(9)32)24(39)45-27-22(38)23-19(35)13(43-27)5-42-25(40)7-3-11(30)17(33)20(36)14(7)15-8(26(41)44-23)4-12(31)18(34)21(15)37/h1-4,13,19,22-23,27-38H,5H2/t13-,19-,22-,23+,27+/m1/s1
Formula Weight 634.0806
Chemical Name or Material Corilagin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.