missing translation for 'onlineSavingsMsg'
Learn More

CORM-401, MedChemExpress

Product Code. 16427047
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16427047 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16427047 Supplier Medchem Express Supplier No. HY109804
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress CORM-401 is an oxidant-sensitive CO-releasing molecule. CORM-401 induces NO increase in the regulation of endothelial calcium signalling. CORM-401 reduces TNF-α/CHX and H2O2-induced ROS production. CORM-401 uncouples mitochondrial respiration and inhibits glycolysis.

CORM-401 is an oxidant-sensitive CO-releasing molecule. CORM-401 induces NO increase in the regulation of endothelial calcium signalling. CORM-401 reduces TNF-α/CHX and H2O2-induced ROS production. CORM-401 uncouples mitochondrial respiration and inhibits glycolysis.

Specifications

Chemical Name or Material CORM-401
Physical Form Solid
CAS 1001015-18-4
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C8H6MnNO6S2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 25 mg/mL (75.48 mM; Need ultrasonic)
SMILES CN(C1=[S][Mn+](C#O)(C#O)(C#O)(C#O)[S-]1)CC([O-])=O.[H+]
Molecular Weight (g/mol) 331.2
Formula Weight 331.2
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.