Learn More
Corning™ 384-Well, Cell Culture-Treated, Flat-Bottom, Low Volume Microplate
Product Code. 10125564 Shop All Corning Products
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 50
Product Code. Color Unit Size
10125564 White Pack of 50
1 options
Product Code. 10125564 Supplier Corning™ Supplier No. 3826
  • ONLINE EXCLUSIVE PRICE
    695.00 €
    648.89 € / Pack of 50
    Save 46.11 €
    6% Off
View all promotions
Estimated Shipment: 05-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Color:White; Sterility:Sterile; Lid:With Lid
Designed to reduce well-to-well cross-talk during fluorescent and luminescent assays.
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Format Standard
Lid With Lid
Culture Area 0.03 cm2
Array 16 x 24
Binding Type TC-Treated
Color White
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 5 to 40 μL
Sterility Sterile
Surface Treatment Cell Culture
Well Shape Flat Bottom (F-well)
For Use With (Application) Provides easier access and imaging for superior assay performance, fluorescence applications, reduce cross-contamination during assay and liquid-handling process
No. per Case 50/Cs.
No. per Pack 10/Pk.
Quantity 50/Cs.
Well Color White