missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 384-Well Nonbinding Surface (NBS™) Microplates
GREENER_CHOICE

Product Code. 10266083 Shop All Corning Products
Change view
Click to view available options
Color:
Black
White
White, Clear Bottom
Well Color:
Black
Clear
White
Barcode:
Includes Barcode
No Barcode
Unit Size:
Pack of 100
Pack of 50
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Well Color Barcode unitSize
10266083 White White Includes Barcode Pack of 50
10728402 Black Clear No Barcode Pack of 100
10317511 White, Clear Bottom Clear No Barcode Pack of 100
10782684 Black Black Includes Barcode Pack of 50
11353725 White Clear Includes Barcode Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10266083 Supplier Corning™ Supplier No. 3574BC
  • ONLINE EXCLUSIVE PRICE
    750.00 €
    699.09 € / Pack of 50
    Save 50.91 €
    6% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Enhanced signal-to-noise ratio

  • Ideal for homogenous assays such as Scintillation Proximity Assays (SPA) and fluorescent assays
  • Modified polymer surface is nonionic hydrophilic, minimizing bio-interaction within wells to reduce nonspecific binding and reagent costs
  • Significantly reduces protein and nucleic acid binding to polymers, maintains enzyme activity, and inhibits adhesion of a number of cell lines Stable to aqueous solutions containing EtOH
  • Use at temperature range of 4° to 37°C (39° to 99°F)
  • Flat-bottom wells
  • Compatible with a wide range of plate readers, liquid handlers, and robotic systems

Generic Bar Coded Plates

  • Code 128 bar code format ensures compatibility with most bar code scanning and software systems
  • To avoid mix-ups, the surface treatment of the microplate wells is easily identified
  • Labels applied to all four sides of the plate No sequence number duplicates

TRUSTED_SUSTAINABILITY

Specifications

Barcode Includes Barcode
Format Standard
Lid Without Lid
Array 16 x 24
Color White
For Use With (Equipment) Plate readers, liquid handlers and robotic systems
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 20 to 80 μL
Sterility Non-sterile
Surface Treatment Non-binding
Well Shape Flat Bottom (F-well)
For Use With (Application) For homogenous assays such as scintillation proximity assays (SPA) and fluorescent assays
No. per Case 50/Cs.
No. per Pack 10/Pk.
Quantity 50/Cs.
Temperature Range (English) 39° to 99°F
Temperature Range (Metric) 4°C to 37°C
Well Color White
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.